Invitrogen
Manufacturer Code:PA584728
Catalog # PIPA584728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC9A3R2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3KARP NHERF-2 SIP-1 TKA-1; Na(+)/H(+) exchange regulatory cofactor NHE-RF2; NHE3 kinase A regulatory protein E3KARP; NHE3 regulatory factor 2; NHERF-2; SIP-1; Sodium-hydrogen exchanger regulatory factor 2; sodium/hydrogen exchanger 3 kinase A regulatory protein; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 2; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 2; solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2; Solute carrier family 9 isoform A3 regulatory factor 2; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 2; SRY-interacting protein 1; TKA-1; Tyrosine kinase activator protein 1
Gene Aliases: E3KARP; NHE3RF2; NHERF-2; NHERF2; OCTS2; SIP-1; SIP1; SLC9A3R2; TKA-1
UniProt ID: (Human) Q15599
Entrez Gene ID: (Human) 9351
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.