Invitrogen
Manufacturer Code:PA521357
Catalog # PIPA521357
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 82 and 307 (Q15599) of NHERF-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3KARP NHE3RF2 NHERF-2 NHERF2 OCTS2 SIP-1 SIP1 TKA-1 NHE3 kinase A regulatory protein E3KARP NHE3 regulatory factor 2 Na(+)/H(+) exchange regulatory cofactor NHE-RF2 SRY-interacting protein 1 sodium/hydrogen exchanger 3 kinase A regulatory protein solute carrier family 9 (sodium/hydrogen exchanger) isoform 3 regulator 2 solute carrier family 9 (sodium/hydrogen exchanger) isoform 3 regulatory factor 2 solute carrier family 9 isoform A3 regulatory factor 2 tyrosine kinase activat; Na(+)/H(+) exchange regulatory cofactor NHE-RF2; NHE3 kinase A regulatory protein E3KARP; NHE3 regulatory factor 2; NHERF-2; Octs2; SIP-1; Sodium-hydrogen exchanger regulatory factor 2; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3 kinase A regulatory protein; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 2; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 2; solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2; solute carrier family 9 isoform 3 regulator 2, isoform A; sodium/hydrogen exchanger; Solute carrier family 9 isoform A3 regulatory factor 2; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 2; Sry interacting protein 1; SRY-interacting protein 1; TKA-1; Tyrosine kinase activator protein 1
Gene Aliases: 0610011L07Rik; 1200011K07Rik; 2010007A20Rik; E3KARP; NHE3RF2; NHERF-2; NHERF2; OCTS2; SIP-1; SIP1; SLC9A3R2; Sryip1; TKA-1
UniProt ID: (Human) Q15599, (Mouse) Q9JHL1
Entrez Gene ID: (Human) 9351, (Mouse) 65962
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.