Novus Biologicals
Manufacturer Code:NBP184918
Catalog # NBP184918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GGDETKLLVVDRETDEFFKKCRVIPSQEHLNGPLPVPFTNGEIQKENSREALAEAALESPRPALVRSASSDTSEELNSQDSPPKQDSTAPSSTSSSDPILDFNISLAMAKERAHQKRSSKRAPQMDWSKKNELFSNL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EBP50; EBP50NHERF-1 ezrin-radixin-moesin binding phosphoprotein-50 Ezrin-radixin-moesin-binding phosphoprotein 50 Na(+)/H(+) exchange regulatory cofactor NHE-RF1 Na+/H+ exchange regulatory co-factor NHERF1solute carrier family 9 (sodium/hydrogen exchanger) isoform 3 regulator 1 NHERFNPHLOP2 Regulatory cofactor of Na(+)/H(+) exchanger sodium/hydrogen exchanger regulatory factor 1 Sodium-hydrogen exchanger regulatory factor 1 solute carrier family 9 (sodium/hydrogen exchanger) isoform 3 regulatoryfactor 1 solute carrier family 9 (sodium/hydrogen exchanger) member 3 regulator 1 Solute carrier family 9 isoform A3 regulatory factor 1; ezrin-radixin-moesin binding phosphoprotein-50; Ezrin-radixin-moesin-binding phosphoprotein 50; Na(+)/H(+) exchange regulatory cofactor NHE-RF1; Na+/H+ exchange regulatory co-factor; NHERF-1; Regulatory cofactor of Na(+)/H(+) exchanger; Sodium-hydrogen exchanger regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 1; Solute carrier family 9 isoform A3 regulatory factor 1; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 1
Gene Aliases: EBP50; NHERF; NHERF-1; NHERF1; NPHLOP2; SLC9A3R1
UniProt ID: (Human) O14745
Entrez Gene ID: (Human) 9368
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.