Invitrogen
Manufacturer Code:PA561400
Catalog # PIPA561400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NHEDC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mitochondrial Na(+)/H(+) exchanger NHA2; Mitochondrial sodium/hydrogen exchanger 9B2 Mitochondrial Na(+)/H(+) exchanger NHA2 Na(+)/H(+) exchanger-like domain-containing protein 2 NHE domain-containing protein 2 Sodium/hydrogen exchanger-like domain-containing protein 2 Solute carrier family 9 subfamily B member 2 SLC9B2 NHA2 NHEDC2; mitochondrial sodium/hydrogen exchanger NHA2; Na(+)/H(+) exchanger NHA2; Na(+)/H(+) exchanger-like domain-containing protein 2; Na+/H+ exchanger domain containing 2; NHE domain-containing protein 2; Sodium/hydrogen exchanger 9B2; Sodium/hydrogen exchanger-like domain-containing protein 2; Solute carrier family 9 subfamily B member 2; solute carrier family 9, subfamily B (cation proton antiporter 2), member 2; solute carrier family 9, subfamily B (NHA2, cation proton antiporter 2), member 2
Gene Aliases: NHA2; NHE10; NHEDC2; SLC9B2
UniProt ID: (Human) Q86UD5
Entrez Gene ID: (Human) 133308
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.