Proteintech
Manufacturer Code:183181AP150UL
Catalog # 50-173-2111
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NHE8 Fusion Protein Ag13111 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0939 Na(+)/H(+) exchanger 8 NHE 8 NHE8 SLC9A8 Sodium/hydrogen exchanger 8; Na(+)/H(+) exchanger 8; NHE-8; Sodium/hydrogen exchanger 8; solute carrier family 9 (sodium/hydrogen exchanger), isoform 8; solute carrier family 9 (sodium/hydrogen exchanger), member 8; Solute carrier family 9 member 8; solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8
Gene Aliases: 1200006P13Rik; 6430709P13Rik; AI182282; KIAA0939; NHE-8; NHE8; SLC9A8
UniProt ID: (Human) Q9Y2E8, (Rat) Q4L208, (Mouse) Q8R4D1
Entrez Gene ID: (Human) 23315, (Rat) 311651, (Mouse) 77031
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.