Novus Biologicals
Manufacturer Code:NBP182574
Catalog # NBP182574
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:DNPVFSPDEALDRSLLARLPPWLSPGETVVPSQRARTQIPYSPGTFCRLMPFRLSSKSVDSFLQADGPEERPPAALPEST |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: isoform 3 MGC126718 NHE3MGC126720 solute carrier family 9 (sodium/hydrogen exchanger) member 3 Solute carrier family 9 member 3; Na(+)/H(+) exchanger 3; NHE-3; Sodium/hydrogen exchanger 3; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3; solute carrier family 9 (sodium/hydrogen exchanger), member 3; Solute carrier family 9 member 3; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3
Gene Aliases: DIAR8; NHE-3; NHE3; SLC9A3
UniProt ID: (Human) P48764
Entrez Gene ID: (Human) 6550
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.