Novus Biologicals
Manufacturer Code:NBP247967UV
Catalog # NBP247967UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NGFR5 + NTR/912 |
Immunogen | NGFR from A875 melanoma cells (NGFR5) Recombinant human p75 NGFR protein (NTR/912) |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CD271; CD271 CD271 antigen Gp80-LNGFR member 16 nerve growth factor receptor NGF receptor p75 ICD TNFRSF16nerve growth factor receptor (TNFR superfamily member 16) tumor necrosis factor receptor superfamily member 16; Gp80-LNGFR; low affinity nerve growth factor receptor; Low affinity neurotrophin receptor p75NTR; Low-affinity nerve growth factor receptor; NGF receptor; p75 ICD; TNFR superfamily, member 16; Tumor necrosis factor receptor superfamily member 16
Gene Aliases: CD271; Gp80-LNGFR; NGFR; p75(NTR); p75NTR; TNFRSF16
UniProt ID: (Human) P08138
Entrez Gene ID: (Human) 4804
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.