Invitrogen
Manufacturer Code:MA513311
Catalog # PIMA513311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NGFR5 |
Immunogen | NGFR from A875 melanoma cells |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD271; Gp80-LNGFR; low affinity nerve growth factor receptor; Low affinity neurotrophin receptor p75NTR; Low-affinity nerve growth factor receptor; Low-affinity nerve growth factor receptor NGF receptor Gp80-LNGFR p75 ICD Low affinity neurotrophin receptor p75NTR p75 LNGFR p75NTR RNNGFRR CD271 p75 p-75; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; p75 ICD; TNFR superfamily, member 16; Tumor necrosis factor receptor superfamily member 16
Gene Aliases: CD271; Gp80-LNGFR; LNGFR; NGFR; p75; p75(NTR); p75NTR; RNNGFRR; TNFRSF16
UniProt ID: (Human) P08138, (Rat) P07174
Entrez Gene ID: (Human) 4804, (Rabbit) 100349095, (Rat) 24596
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.