Invitrogen
Manufacturer Code:MA529736
Catalog # PIMA529736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 117 |
Immunogen | Recombit Mouse NGFR/P75 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD271; Low affinity neurotrophin receptor p75NTR; Low-affinity nerve growth factor receptor; Low-affinity nerve growth factor receptor NGF receptor Gp80-LNGFR p75 ICD Low affinity neurotrophin receptor p75NTR p75 LNGFR p75NTR RNNGFRR CD271 p75 p-75; NGF receptor; p75 neurotrophin receptor; Tumor necrosis factor receptor superfamily member 16
Gene Aliases: LNGFR; Ngfr; p75; p75NGFR; p75NTR; Tnfrsf16
UniProt ID: (Mouse) Q9Z0W1
Entrez Gene ID: (Mouse) 18053
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.