Novus Biologicals
Manufacturer Code:NBP219613
Catalog # NBP219613
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human NUR77. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Early response protein NAK1; Early response protein NAK1 GFRP1ST-59 growth factor-inducible nuclear protein N10 HMRNP10 hormone receptor MGC9485 N10 NAK1 NAK-1 NGFIB Nuclear hormone receptor NUR/77 nuclear receptor subfamily 4 group A member 1 nuclear receptor subfamily 4 group A member 1 NUR77 Orphan nuclear receptor HMR Orphan nuclear receptor TR3 steroid receptor TR3 Testicular receptor 3 TR3 TR3 orphan receptor; growth factor-inducible nuclear protein N10; hormone receptor; nerve growth factor IB nuclear receptor variant 1; Nuclear hormone receptor NUR/77; Nuclear receptor subfamily 4 group A member 1; nuclear receptor subfamily 4, group A, member 1; Nur77; Orphan nuclear receptor HMR; Orphan nuclear receptor TR3; ST-59; steroid receptor TR3; Testicular receptor 3; TR3 orphan receptor
Gene Aliases: GFRP1; HMR; N10; NAK-1; NAK1; NGFIB; NP10; NR4A1; NUR77; TR3
UniProt ID: (Human) P22736
Entrez Gene ID: (Human) 3164
Molecular Function: nuclear hormone receptor receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.