Invitrogen
Manufacturer Code:MA112347
Catalog # 01-674-286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AS21 |
Immunogen | Recombit protein corresponding to residues surrounding aa 46-62 of human NGF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-nerve growth factor; Beta-NGF; Nerve Growth Factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit
Gene Aliases: Beta-NGF; HSAN5; NGF; NGFB
UniProt ID: (Bovine) P13600, (Human) P01138, (Mouse) P01139
Entrez Gene ID: (Bovine) 281350, (Human) 4803, (Mouse) 18049
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.