Invitrogen
Manufacturer Code:PA579591
Catalog # PIPA579591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat Lipocalin 2 recombit protein (Position: Q21-N198). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 24p3; Alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin-2; Neutrophil gelatinase-associated lipocalin; Neutrophil gelatinase-associated lipocalin NGAL Alpha-2-microglobulin-related protein Alpha-2U globulin-related protein Lipocalin-2 Siderocalin LCN2 p25 Lcn2; NGAL; Oncogene 24p3; p25; secreted inducible protein 24; Siderocalin LCN2; SV-40-induced 24P3 protein
Gene Aliases: 24p3; AW212229; Lcn2; Sip24
UniProt ID: (Mouse) P11672, (Rat) P30152
Entrez Gene ID: (Mouse) 16819, (Rat) 170496
Molecular Function:
isomerase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.