R&D Systems
Manufacturer Code:AF2585
Catalog # AF2585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human NG2/MCSP Ser1583-Ser2224 Accession # Q6UVK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AN2 chondroitin sulfate proteoglycan 4 chondroitin sulfate proteoglycan 4 (melanoma-associated) CSPG4 EC 3.6.3 HMW-MAA MCSP MCSPChondroitin sulfate proteoglycan NG2 MCSPG MCSPGMelanoma chondroitin sulfate proteoglycan MEL-CSPG MSK16Melanoma-associated chondroitin sulfate proteoglycan NG2EC 2.7.8; Chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); Chondroitin sulfate proteoglycan NG2; Melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan
Gene Aliases: CSPG4; HMW-MAA; MCSP; MCSPG; MEL-CSPG; MSK16; NG2
UniProt ID: (Human) Q6UVK1
Entrez Gene ID: (Human) 1464
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.