Novus Biologicals
Manufacturer Code:NBP267356
Catalog # NBP267356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SZ20-01 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: DKFZp686C01211 DNA binding factor KBF1 DNA-binding factor KBF1 EBP-1 KBF1 NF-kappaB NF-kappa-B NF-kappabeta NFKB-p105 NFKB-p50 nuclear factor kappa-B DNA binding subunit nuclear factor NF-kappa-B p105 subunit nuclear factor NF-kappa-B p50 subunit nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151 p105 p50; DNA-binding factor KBF1; EBP-1; NF-kappabeta; nuclear factor kappa-B DNA binding subunit; Nuclear factor NF-kappa-B p105 subunit; Nuclear factor NF-kappa-B p50 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
Gene Aliases: CVID12; EBP-1; KBF1; NF-kappa-B; NF-kappaB; NF-kB1; NFkappaB; NFKB-p105; NFKB-p50; NFKB1; p105; p50
UniProt ID: (Human) P19838
Entrez Gene ID: (Human) 4790
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.