Novus Biologicals
Manufacturer Code:NBP177395B
Catalog # NBP177395B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A genomic peptide made to an N-terminal region of the human NFkB p105/p50 protein (within residues 400-590). [Swiss-Prot P19838] |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-binding factor KBF1; DNA-binding factor KBF1 EBP-1 NF-kappa-B NF-kB1 NFKB1 NFKB-p105 Nuclear factor NF-kappa-B p105 subunit Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1 p105; EBP-1; NF-kappabeta; nuclear factor kappa-B DNA binding subunit; Nuclear factor NF-kappa-B p105 subunit; Nuclear factor NF-kappa-B p50 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
Gene Aliases: CVID12; EBP-1; KBF1; NF-kappa-B; NF-kappaB; NF-kB1; NFkappaB; NFKB-p105; NFKB-p50; NFKB1; p105; p50
UniProt ID: (Human) P19838
Entrez Gene ID: (Human) 4790
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.