Invitrogen
Manufacturer Code:PA130408
Catalog # PIPA130408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide EDFSSIADMDFSALLSQISS corresponding to the C-terminus of Human NFkB p65. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: avian reticuloendotheliosis viral (v-rel) oncogene homolog A; nf&questb; NF-kappa-B p65delta3; NF-kappa-B transcription factor p65; nuclear factor kappa B subunit p65; Nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; p65 NF kappaB; p65 NF-kappa B; p65 NFkB; Transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A; v-rel reticuloendotheliosis viral oncogene homolog A
Gene Aliases: NFkB; NFKB3; p65; RELA
UniProt ID: (Human) Q04206, (Mouse) Q04207
Entrez Gene ID: (Human) 5970, (Mouse) 19697, (Rat) 309165
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.