Invitrogen
Manufacturer Code:MA526740
Catalog # PIMA526740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-299 of NFS1 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cysteine desulfurase, mitochondrial; NFS1 nitrogen fixation 1 homolog; NFS1 nitrogen fixation 1 homolog (S. cerevisiae) IscS NIFS HUSSY-08; nitrogen fixation 1 (S. cerevisiae, homolog); nitrogen-fixing bacteria S-like protein
Gene Aliases: HUSSY-08; IscS; NFS1; NIFS
UniProt ID: (Human) Q9Y697
Entrez Gene ID: (Human) 9054
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.