Invitrogen
Manufacturer Code:PA576984
Catalog # PIPA576984
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human NFS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cysteine desulfurase mitochondrial NFS1 NIFS HUSSY-08; Cysteine desulfurase, mitochondrial; m-Nfs1; NFS1 nitrogen fixation 1 homolog; nifS-like (sic); nitrogen fixation 1 (S. cerevisiae, homolog); nitrogen fixation gene 1; nitrogen fixation gene, yeast homolog 1; nitrogen-fixing bacteria S-like protein
Gene Aliases: AA987187; HUSSY-08; IscS; m-Nfs1; m-Nfsl; NFS1; NIFS
UniProt ID: (Human) Q9Y697, (Mouse) Q9Z1J3
Entrez Gene ID: (Human) 9054, (Rat) 84594, (Mouse) 18041
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.