Invitrogen
Manufacturer Code:PA540653
Catalog # PIPA540653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human NFS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cysteine desulfurase, mitochondrial; Cysteine desulfurase, mitochondrial-like protein; m-Nfs1; NFS1 nitrogen fixation 1 homolog; NFS1 nitrogen fixation 1 homolog (S. cerevisiae) IscS NIFS HUSSY-08; nifS-like (sic); nitrogen fixation 1 (S. cerevisiae, homolog); nitrogen fixation gene 1; nitrogen fixation gene, yeast homolog 1; nitrogen-fixing bacteria S-like protein
Gene Aliases: AA987187; HUSSY-08; IscS; m-Nfs1; m-Nfsl; NFS1; NIFS
UniProt ID: (Human) Q9Y697, (Mouse) Q9Z1J3
Entrez Gene ID: (Bovine) 517656, (Horse) 100054930, (Human) 9054, (Dog) 477214, (Rabbit) 100328709, (Rat) 84594, (Mouse) 18041, (Guinea pig) 100733215
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.