Novus Biologicals
Manufacturer Code:NBP243742
Catalog # NBP243742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 377 |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human NFKBIL2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: I-kappa-B-related protein; ikappaBR; IkappaBR I-kappa-B-related protein IKBRFLJ40087 Inhibitor of kappa B-related protein NF-kappa-B inhibitor-like protein 2 NFKBIL2tonsoku-like protein Nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 2ikappaBR tonsoku-like DNA repair protein; Inhibitor of kappa B-related protein; NF-kappa-B inhibitor-like protein 2; Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 2; Tonsoku-like protein
Gene Aliases: IKBR; NFKBIL2; TONSL
UniProt ID: (Human) Q96HA7
Entrez Gene ID: (Human) 4796
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.