Invitrogen
Manufacturer Code:PA571752
Catalog # PIPA571752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 327-356 amino acids from the C-terminal region of human NFKBIL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: I-kappa-B-like protein; ikappaBL; Inhibitor of kappa B-like protein; NF-kappa-B inhibitor-like protein 1; NF-kappa-B inhibitor-like protein 1 Inhibitor of kappa B-like protein I-kappa-B-like protein IkappaBL Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1 NFKBIL1 IKBL; Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1
Gene Aliases: IKBL; LST1; NFKBIL; NFKBIL1
UniProt ID: (Human) Q9UBC1
Entrez Gene ID: (Human) 4795
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.