Novus Biologicals
Manufacturer Code:NBP170344
Catalog # NBP170344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Amino acids RRYLRVQQVRWHPDRFLQR (331-349) of human NFKBIL1 were used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: I-kappa-B-like protein; ikappaBL; IkappaBL I-kappa-B-like protein IKBLikappaBL Inhibitor of kappa B-like protein LST1 NF-kappa-B inhibitor-like protein 1 nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 1NFKBIL; Inhibitor of kappa B-like protein; NF-kappa-B inhibitor-like protein 1; Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1
Gene Aliases: IKBL; LST1; NFKBIL; NFKBIL1
UniProt ID: (Human) Q9UBC1
Entrez Gene ID: (Human) 4795
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.