Invitrogen
Manufacturer Code:PA571595
Catalog # PIPA571595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 91-119 amino acids from the N-terminal region of human NFKBID |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: I-kappa-B-delta; ikappaBdelta; IkappaBNS; IkB-delta; NF-kappa B inhibitor; NF-kappa-B inhibitor delta; NF-kappa-B inhibitor delta I-kappa-B-delta IkB-delta IkappaBdelta IkappaBNS T-cell activation NFKB-like protein TA-NFKBH NFKBID IKBNS; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, delta
Gene Aliases: IkappaBNS; Ikbns; Nfkbid
UniProt ID: (Mouse) Q2TB02
Entrez Gene ID: (Mouse) 243910
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.