Novus Biologicals
Manufacturer Code:NBP305193
Catalog # NB093532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human ARHGAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: delta IkappaBdelta I-kappa-B-delta IkappaBNSikappaBdelta ikB-delta IKBNS MGC11314 MGC149503 nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor TA-NFKBHNF-kappa-B inhibitor delta T-cell activation NFKB-like protein; I-kappa-B-delta; ikappaBdelta; IkappaBNS; IkB-delta; NF-kappa-B inhibitor delta; NFKB inhibitor delta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, delta; T-cell activation NFKB-like protein; TA-NFKBH
Gene Aliases: IkappaBNS; IKBNS; NFKBID; TA-NFKBH
UniProt ID: (Human) Q8NI38
Entrez Gene ID: (Human) 84807
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.