Abnova
Manufacturer Code:PAB25387
Catalog # 89-127-023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding S32/36 of human NFKBIA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of NFKBIA.
For Research Use Only
Protein Aliases: I-kappa-B-alpha; IkappaBalpha; IkB-alpha; IKBA; MAD-3; Major histocompatibility complex enhancer-binding protein MAD3; NF-kappa-B inhibitor alpha; NFKBI; nuclear factor of kappa light chain gene enhancer in B-cells; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha
Gene Aliases: IKBA; MAD-3; MAD3; NFKBI; NFKBIA
UniProt ID: (Human) P25963
Entrez Gene ID: (Human) 4792
Molecular Function:
enzyme modulator
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.