Invitrogen
Manufacturer Code:PA579736
Catalog # PIPA579736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human NFIB/NF1B2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCAAT-box-binding transcription factor; CTF; NF-I/B; NF1-B; Nuclear factor 1 B-type; Nuclear factor 1 B-type NF1-B Nuclear factor 1/B CCAAT-box-binding transcription factor CTF Nuclear factor I/B NF-I/B NFI-B TGGCA-binding protein NFIB; nuclear factor 1/B; Nuclear factor I/B; olfactory epithelium nuclear factor I-B; TGGCA-binding protein
Gene Aliases: 6720429L07Rik; CTF; E030026I10Rik; HMGIC/NFIB; NF-I/B; NF1-B; NFI-B; NFI-RED; NFIB; NFIB2; NFIB3
UniProt ID: (Human) O00712, (Mouse) P97863
Entrez Gene ID: (Human) 4781, (Mouse) 18028, (Rat) 29227
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.