Novus Biologicals
Manufacturer Code:NBP181406
Catalog # NBP181406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VKSPQCSNPGLCVQPHHIGVSVKELDLYLAYFVHAADSSQSESPSQPSDADIKDQPENGHLGFQDSFVTSGVFSVTELVRVSQTPIAAGTGPNFSLSDLESSSYYSMSPGA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCAAT-box-binding transcription factor; CCAAT-box-binding transcription factor CTF DKFZp686J23256 FLJ39164 KIAA1439DKFZp434L0422 NF1-A NF-I/A NFI-A NFI-L nuclear factor 1 A-type Nuclear factor 1/A nuclear factor I/ATGGCA-binding protein; CTF; NF-I/A; NF1-A; Nuclear factor 1 A-type; Nuclear factor I/A; TGGCA-binding protein
Gene Aliases: CTF; KIAA1439; NF-I/A; NF1-A; NFI-A; NFI-L; NFIA
UniProt ID: (Human) Q12857
Entrez Gene ID: (Human) 4774
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.