Invitrogen
Manufacturer Code:PA564222
Catalog # PIPA564222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NFE2L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endoplasmic reticulum membrane sensor NFE2L1; LCR-F1; LCR-F1 NRF1 TCF11 NF-E2-related factor 1 NFE2-related factor 1 TCF-11 locus control region-factor 1 nuclear factor erythroid 2-related factor 1 nuclear factor erythroid derived 2 like 1 transcription factor 11 (basic leucine zipper type) transcription factor HBZ17 transc; Locus control region-factor 1; NF-E2-related factor 1; NFE2-related factor 1; nuclear factor (erythroid-derived 2)-like 1; Nuclear factor erythroid 2-related factor 1; nuclear factor, erythroid 2-like 1; Nuclear factor, erythroid derived 2, like 1; Protein NRF1, p110 form; Protein NRF1, p120 form; TCF-11; Transcription factor 11; transcription factor 11 (basic leucine zipper type); transcription factor HBZ17; transcription factor LCR-F1; Transcription factor NRF1
Gene Aliases: HBZ17; LCR-F1; NFE2L1; NRF1; TCF11
UniProt ID: (Human) Q14494
Entrez Gene ID: (Human) 4779
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
nucleic acid binding
protease
serine protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.