Abnova
Manufacturer Code:H00004779A01
Catalog # 89-104-988
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 677 a.a. &bsim 771 a.a) partial recombinant protein with GST tag.,NFE2L1 (NP_003195 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant NFE2L1.
For Research Use Only
Protein Aliases: Endoplasmic reticulum membrane sensor NFE2L1; FLJ00380; LCR-F1; Locus control region-factor 1; NF-E2-related factor 1; NFE2-related factor 1; NRF1; nuclear factor (erythroid-derived 2)-like 1; Nuclear factor erythroid 2-related factor 1; nuclear factor, erythroid 2-like 1; Nuclear factor, erythroid derived 2, like 1; Protein NRF1, p110 form; Protein NRF1, p120 form; TCF-11; TCF11; Transcription factor 11; transcription factor 11 (basic leucine zipper type); transcription factor HBZ17; transcription factor LCR-F1; Transcription factor NRF1
Gene Aliases: HBZ17; LCR-F1; NFE2L1; NRF1; TCF11
UniProt ID: (Human) Q14494
Entrez Gene ID: (Human) 4779
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
nucleic acid binding
protease
serine protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.