Proteintech
Manufacturer Code:664361IG150UL
Catalog # 50-173-6862
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D7C8 |
Immunogen | NFE2 Fusion Protein Ag1570 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Leucine zipper protein NF E2 NF E2 NFE2 p45 p45 NF E2; Leucine zipper protein NF-E2; nuclear factor (erythroid-derived 2), 45kD; nuclear factor (erythroid-derived 2), 45kDa; Nuclear factor, erythroid-derived 2 45 kDa subunit; p45 NF-E2; Transcription factor NF-E2 45 kDa subunit
Gene Aliases: NF-E2; NFE2; p45
UniProt ID: (Human) Q16621
Entrez Gene ID: (Human) 4778
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
nucleic acid binding
protease
serine protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.