Invitrogen
Manufacturer Code:PA527762
Catalog # PIPA527762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 373 of Human NF-E2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Leucine zipper protein NF-E2; NF-E2 p45 leucine zipper protein NF-E2 nuclear factor erythroid-derived 2 45 kDa subunit p45 NF-E2 transcription factor NF-E2 45 kDa subunit; nuclear factor (erythroid-derived 2), 45kD; nuclear factor (erythroid-derived 2), 45kDa; Nuclear factor, erythroid-derived 2 45 kDa subunit; p45 NF-E2; Transcription factor NF-E2 45 kDa subunit
Gene Aliases: NF-E2; NF-E2/P45; NFE2; p45; p45nf-e2; p45NFE2
UniProt ID: (Human) Q16621, (Mouse) Q07279
Entrez Gene ID: (Human) 4778, (Mouse) 18022
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
nucleic acid binding
protease
serine protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.