Invitrogen
Manufacturer Code:PA5100881
Catalog # PIPA5100881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NFE2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Leucine zipper protein NF-E2; leucine zipper protein NF-E2 NFE2 NF-E2 NF-E2/P45 nuclear factor (erythroid-derived 2) 45kD nuclear factor (erythroid-derived 2) 45kDa nuclear factor erythroid 2 nuclear factor erythroid derived 2 nuclear factor erythroid-derived 2 45 kDa subunit p45 p45 NF-E2 p45NFE2 p45nf-e2 Transcription factor NF-E2 45 kDa subunit; nuclear factor (erythroid-derived 2), 45kD; nuclear factor (erythroid-derived 2), 45kDa; nuclear factor, erythroid derived 2; Nuclear factor, erythroid-derived 2 45 kDa subunit; p45 NF-E2; Transcription factor NF-E2 45 kDa subunit
Gene Aliases: NF-E2; NF-E2/P45; NFE2; p45; p45nf-e2; p45NFE2
UniProt ID: (Human) Q16621, (Rat) Q6AYT2, (Mouse) Q07279
Entrez Gene ID: (Human) 4778, (Rat) 366998, (Mouse) 18022
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
nucleic acid binding
protease
serine protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.