Invitrogen
Manufacturer Code:PA123423
Catalog # PIPA123423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding top residues L(102) S S G H T R P D G A P A(114) of human NFATc1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NF-ATc; NF-ATc1; NFAT transcription complex cytosolic component; NFATC1 nuclear factor of activated T-cells cytoplasmic calcineurin-dependent 1 MGC138448 NFAT2 NFATc NFATC NF-ATc NF-ATC NF-ATc1 NFAT trans; nuclear factor of activated T-cells 'c'; Nuclear factor of activated T-cells, cytoplasmic 1; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1
Gene Aliases: NF-ATC; NF-ATc1.2; NFAT2; NFATC; NFATC1
UniProt ID: (Human) O95644
Entrez Gene ID: (Human) 4772
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.