Invitrogen
Manufacturer Code:PA599546
Catalog # PIPA599546
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NFAT4 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C80703 D8Ertd281e NFAC3 NFAT4 NF-AT4 NFATC3 NF-ATc3 NFATX NFC3 nuclear factor of activated T cells cytoplasmic calcineurin dependent 3 nuclear factor of activated T-cells 3 nuclear factor of activated T-cells c3 isoform IE-Xa nuclear factor of activated T-cells cytoplasmic 3 nuclear factor of activated T-cells cytoplasmic calcineurin-dependent 3 T cell transcription factor NFAT4 T-cell transcription factor NFAT4; NF-AT4; NF-ATc3; NFATx; nuclear factor of activated T-cells c3 isoform IE-Xa; Nuclear factor of activated T-cells, cytoplasmic 3; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3; T cell transcription factor NFAT4; T-cell transcription factor NFAT4
Gene Aliases: C80703; D8Ertd281e; NFAT4; NFATC3; NFATX
UniProt ID: (Human) Q12968
Entrez Gene ID: (Human) 4775, (Mouse) 18021
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.