Invitrogen
Manufacturer Code:PA578403
Catalog # PIPA578403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of mouse NFAT4. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NF-AT4; NF-ATc3; Nfatc3 nuclear factor of activated T cells cytoplasmic calcineurin dependent 3; nuclear factor of activated T-cells, cytoplasmic 3; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3; T-cell transcription factor NFAT4
Gene Aliases: C80703; D8Ertd281e; NFAT4; NFATx
Entrez Gene ID: (Mouse) 18021
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.