Novus Biologicals
Manufacturer Code:NB300504AF350
Catalog # NB30504A350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 25A10.D6.D2 |
Immunogen | Synthetic peptide corresponding to residues A(51) I S S P S G L A Y P D D V L D Y G L(69) of mouse NFATc2-A B and C isoforms. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: NF-ATc2; NF-ATp; NFAT pre-existing subunit; NFAT pre-existing subunit NFAT transcription complex preexisting component NFAT1nuclear factor of activated T-cells cytoplasmic 2 NFATc2 NF-ATc2 NFATp NF-ATp NFATPnuclear factor of activated T-cells preexisting component nuclear factor of activated T-cells cytoplasmic calcineurin-dependent 2 preexisting nuclear factor of activated T-cells 2 T cell transcription factor NFAT1 T-cell transcription factor NFAT1; NFAT transcription complex, preexisting component; Nuclear factor of activated T-cells, cytoplasmic 2; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2; nuclear factor of activated T-cells, preexisting component; preexisting nuclear factor of activated T-cells 2; T cell transcription factor NFAT1; T-cell transcription factor NFAT1
Gene Aliases: NFAT1; NFATC2; NFATP
UniProt ID: (Human) Q13469
Entrez Gene ID: (Human) 4773
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.