Invitrogen
Manufacturer Code:PA519164
Catalog # PIPA519164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QQFRTGLSSPDARYQ) corresponding to the internal amino acids of NFATC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: NF-ATc2; NF-ATp; NFAT pre-existing subunit; NFAT transcription complex, preexisting component; NFATC2 nuclear factor of activated T-cells cytoplasmic calcineurin-dependent 2 KIAA0611 NFAT1 NFATP NFAT pre-existing subunit NFAT transcription complex preexisting component OTTHUMP00000031291 T cell transcription factor NFAT1 nuclear factor of activated T-cells cytoplasmic 2 nuclear factor of activated T-cells preexisting component preexisting nuclear factor of activated T-cells 2; Nuclear factor of activated T-cells, cytoplasmic 2; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2; nuclear factor of activated T-cells, preexisting component; preexisting nuclear factor of activated T-cells 2; T cell transcription factor NFAT1; T-cell transcription factor NFAT1
Gene Aliases: NFAT1; NFATC2; NFATP
UniProt ID: (Human) Q13469
Entrez Gene ID: (Human) 4773
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.