Proteintech
Manufacturer Code:220231AP150UL
Catalog # 50-173-3689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NFATC2 Fusion Protein Ag16849 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NF ATc2 NF ATp NFAT pre existing subunit NFAT1 NFATC2 NFATP; NF-ATc2; NF-ATp; NFAT pre-existing subunit; NFAT transcription complex, preexisting component; Nuclear factor of activated T-cells, cytoplasmic 2; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2; nuclear factor of activated T-cells, preexisting component; preexisting nuclear factor of activated T-cells 2; T cell transcription factor NFAT1; T-cell transcription factor NFAT1
Gene Aliases: AI607462; NF-ATc2; NF-ATp; NFAT1; NFAT1-D; NFATC2; NFATP
UniProt ID: (Human) Q13469, (Mouse) Q60591
Entrez Gene ID: (Human) 4773, (Mouse) 18019
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.