Novus Biologicals
Manufacturer Code:NB10056732
Catalog # NB10056732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Amino acids 264-282 (NKRKYSLNGRQPPYSPHHS) of human NFATc1 were used as immunogen for this antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC138448 NFAT transcription complex cytosolic component NFAT2cytoplasmic 1 NFATC NF-ATC NFATc1 NF-ATc1 nuclear factor of activated T-cells cytoplasmic calcineurin-dependent 1; NF-ATc; NF-ATc1; NFAT transcription complex cytosolic component; nuclear factor of activated T-cells 'c'; Nuclear factor of activated T-cells, cytoplasmic 1; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1
Gene Aliases: NF-ATC; NF-ATc1.2; NFAT2; NFATC; NFATC1
UniProt ID: (Human) O95644
Entrez Gene ID: (Human) 4772
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.