Novus Biologicals
Manufacturer Code:NB1203446
Catalog # NB1203446
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Gel Shift (GS) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues C D(1439) L L V S L Q N Q G N N L T G S F(1455) of human NFAT5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.6.1 EC 3.6.3.14 KIAA0827glutamine rich protein H65 NF-AT5NFAT-like protein 1 NFATL1 NFATZ nuclear factor of activated T-cells 5 tonicity-responsive OREBP osmotic response element-binding protein T-cell transcription factor NFAT5 TonE-binding protein TonEBP TONEBPnuclear factor of activated T-cells 5 Tonicity-responsive enhancer-binding protein; glutamine rich protein H65; NF-AT5; NFAT-like protein 1; Nuclear factor of activated T-cells 5; osmotic response element-binding protein; T-cell transcription factor NFAT5; TonE-binding protein; Tonicity-responsive enhancer-binding protein
Gene Aliases: KIAA0827; NF-AT5; NFAT5; NFATL1; NFATZ; OREBP; TONEBP
UniProt ID: (Human) O94916
Entrez Gene ID: (Human) 10725
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.