Abnova
Manufacturer Code:PAB0643
Catalog # 89-114-321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to amino acids 250-265 of human NFAM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Protein Aliases: Calcineurin/NFAT-activating ITAM-containing protein; NFAT activation molecule 1; NFAT-activating protein with ITAM motif 1
Gene Aliases: CNAIP; NFAM1
UniProt ID: (Human) Q8NET5
Entrez Gene ID: (Human) 150372
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.