Novus Biologicals
Manufacturer Code:NBP303740100UL
Catalog # NB092633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 477-576 of human NF2/Merlin (NP_000259.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACN BANF merlin moesin-ezrin-radixin like Moesin-ezrin-radixin-like protein moesin-ezrin-radizin-like protein neurofibromin 2 (bilateral acoustic neuroma) neurofibromin 2 (merlin) neurofibromin-2 SCH Schwannomerlin Schwannomin; Merlin; moesin-ezrin-radixin like; Moesin-ezrin-radixin-like protein; moesin-ezrin-radizin-like protein; neurofibromin 2 (bilateral acoustic neuroma); neurofibromin 2 (merlin); Neurofibromin-2; Schwannomerlin; Schwannomin
Gene Aliases: ACN; BANF; NF2; SCH
UniProt ID: (Human) P35240
Entrez Gene ID: (Human) 4771
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.