Invitrogen
Manufacturer Code:MA1084
Catalog # 01-670-103
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Affinity Purification (AP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | McNFn27a |
Immunogen | Synthetic peptide corresponding to the N-terminal residues 27-41 of human neurofibromin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: neurofibromatosis 1; Neurofibromatosis type 1; Neurofibromatosis-related protein NF-1; Neurofibromin; Neurofibromin truncated; NF1 WSS NFNS VRNF
Gene Aliases: AW494271; E030030H24Rik; Nf-1; NF1; NFNS; VRNF; WSS
UniProt ID: (Human) P21359, (Mouse) Q04690, (Rat) P97526
Entrez Gene ID: (Human) 4763, (Mouse) 18015, (Rat) 24592
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.