Invitrogen
Manufacturer Code:PA146246
Catalog # PIPA146246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | N-terminal peptide a.a. 27-41. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686J1293 FLJ21220 NFNS VRNF WSS; neurofibromatosis 1; Neurofibromatosis type 1; Neurofibromatosis-related protein NF-1; Neurofibromin; Neurofibromin truncated
Gene Aliases: AW494271; E030030H24Rik; Nf-1; NF1; NFNS; VRNF; WSS
UniProt ID: (Human) P21359, (Mouse) Q04690, (Rat) P97526
Entrez Gene ID: (Human) 4763, (Mouse) 18015, (Rat) 24592
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.