Abnova
Manufacturer Code:H00004763A01
Catalog # 89-104-986
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 2719 a.a. &bsim 2818 a.a) partial recombinant protein with GST tag.,NF1 (NP_000258 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant NF1.
For Research Use Only
Protein Aliases: DKFZp686J1293; FLJ21220; Neurofibromatosis-related protein NF-1; Neurofibromin; Neurofibromin truncated; NFNS; VRNF; WSS
Gene Aliases: NF1; NFNS; VRNF; WSS
UniProt ID: (Human) P21359
Entrez Gene ID: (Human) 4763
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.