Novus Biologicals
Manufacturer Code:NBP1052190.025ML
Catalog # NB018362
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry - Free Floating (IHC (Free)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant human NF-L protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: 68 kDa neurofilament protein; 68 kDa neurofilament protein neurofilament protein light chain neurofilament subunit NF-L Neurofilament triplet L protein neurofilament light polypeptide neurofilament-light NF68FLJ53642 NF-L NFLlight polypeptide 68kDa; light molecular weight neurofilament protein; Neurofilament light polypeptide; neurofilament protein, light chain; neurofilament subunit NF-L; Neurofilament triplet L protein; neurofilament, light polypeptide 68kDa; NF-L; protein phosphatase 1, regulatory subunit 110
Gene Aliases: CMT1F; CMT2E; NEFL; NF-L; NF68; NFL; PPP1R110
UniProt ID: (Human) P07196
Entrez Gene ID: (Human) 4747
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.