Invitrogen
Manufacturer Code:MA119172
Catalog # 01-675-942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NF-01 |
Immunogen | Pellet of pig brain cold-stable proteins after depolymerization of microtubules. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 200 kDa neurofilament protein; neurofilament 200kDa; Neurofilament heavy polypeptide; Neurofilament triplet H protein; neurofilament, heavy polypeptide 200kDa; NF-H; NFH NF-H NHC 200 kDa neurofilament protein KIAA0845 Neurofilament heavy polypeptide Neurofilament triplet H protein
Gene Aliases: CMT2CC; KIAA0845; mKIAA0845; NEFH; NF-H; NF200; NFH
UniProt ID: (Human) P12036, (Mouse) P19246
Entrez Gene ID: (Human) 4744, (Pig) 100156492, (Rat) 24587, (Mouse) 380684
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.