Novus Biologicals
Manufacturer Code:NBP1052100.025ML
Catalog # NB018360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry - Free Floating (IHC (Free)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9B12 |
Immunogen | Native NF-H purified from bovine spinal cord binding to phosphorylated KSP sequences. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 200 kDa neurofilament protein; 200 kDa neurofilament protein KIAA0845 Neurofilament Heavy (200kDa) neurofilament heavy polypeptide Neurofilament triplet H protein neurofilament heavy polypeptide neurofilament heavy polypeptide 200kDa NF200 NFH NF-H; Neurofilament heavy polypeptide; Neurofilament triplet H protein; neurofilament, heavy polypeptide 200kDa; NF-H
Gene Aliases: CMT2CC; KIAA0845; NEFH; NFH
UniProt ID: (Human) P12036
Entrez Gene ID: (Human) 4744
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.