Invitrogen
Manufacturer Code:PA597057
Catalog # PIPA597057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 821-1020 of human NEFH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 200 kDa neurofilament protein; neurofilament 200kDa; Neurofilament heavy polypeptide; Neurofilament triplet H protein; NF-H
Gene Aliases: Kiaa0845; mKIAA0845; Nefh; NF-H; NF200; Nfh
UniProt ID: (Mouse) P19246
Entrez Gene ID: (Rat) 24587, (Mouse) 380684
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.