Invitrogen
Manufacturer Code:PA110002
Catalog # 01-670-259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Purified axonal form of bovine NF-H. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: 200 kDa neurofilament protein; heavy neurofilament protein; intermediate filament protein; neurofilament 200kDa; Neurofilament heavy polypeptide; Neurofilament triplet H protein; neurofilament, heavy polypeptide 200kDa; NF-H; NFH NHC NF 200 kD
Gene Aliases: CMT2CC; KIAA0845; mKIAA0845; NEFH; NF-H; NF200; NFH
UniProt ID: (Human) P12036, (Mouse) P19246
Entrez Gene ID: (Dog) 442940, (Horse) 100064328, (Human) 4744, (Pig) 100156492, (Rat) 24587, (Mouse) 380684
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.